Learn More
Biomatik Corporation Rat INSR-β (Insulin Receptor Beta Subunit) ELISA Kit
Supplier:  Biomatik Corporation EKF59134
Encompass_Preferred
Rat INSR-β (Insulin Receptor Beta Subunit) ELISA Kit for detecting INSR-β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.938-60ng/ml. Sensitivity: 0.563ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15127. Target Synonyms: INSR-β. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6331