Learn More
Biomatik Corporation Rat BNP (Brain Natriuretic Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF59148
Encompass_Preferred
Rat BNP (Brain Natriuretic Peptide) ELISA Kit for detecting BNP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13205. Target Synonyms: BNP; Brain Natriuretic Peptide; NPPB; GC-B; B-Type Natriuretic Peptide; Natriuretic Peptide Precursor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6343