Learn More
Biomatik Corporation Mouse Fabp7 (Fatty acid-binding protein, brain) ELISA Kit
Supplier:  Biomatik Corporation EKF59165
Encompass_Preferred
Mouse Fabp7 (Fatty acid-binding protein, brain) ELISA Kit for detecting Fabp7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51880. Target Synonyms: Fabp7; FABP7; B-FABP; BLBP; B-FABPDKFZp547J2313; BLBPMRG; Brain lipid-binding protein; Brain-type fatty acid-binding protein; FABPBbrain lipid binding protein; Fatty acid-binding protein 7; fatty acid-binding protein; brain; Mammary-derived growth inhibitor related; mammary-derived growth inhibitor-related. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6357