Learn More
Biomatik Corporation Mouse MUC1 (Mucin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59181
Encompass_Preferred
Mouse MUC1 (Mucin-1) ELISA Kit for detecting MUC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02496. Target Synonyms: MUC1; CD227; Episialin; H23AG; KL-6; PEM; PEMT; Breast carcinoma-associated antigen DF3; Carcinoma-associated mucin; CD227; CD227 antigen; DF3 antigen; EMA; episialin; H23 antigen; H23AG; KL-6; MAM6; MUC1; ZD; mucin 1; cell surface associated; mucin 1; transmembrane; mucin-1; Peanut-reactive urinary mucin; PEMMUC-1; SEC; PEMT; Polymorphic epithelial mucin; PUMMUC-1; X; tumor associated epithelial mucin; Tumor-associated epithelial membrane antigen; Tumor-associated mucin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6368