Learn More
Biomatik Corporation Human Acyl-CoA desaturase (delta-9-desaturase) ELISA Kit
Supplier:  Biomatik Corporation EKF59192
Encompass_Preferred
Human Acyl-CoA desaturase (delta-9-desaturase) ELISA Kit for detecting Acyl-CoA desaturase in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00767. Target Synonyms: Acyl CoA desaturase; Delta; 9 desaturase; FADS5; Fatty acid desaturase; MSTP008; SCD; SCD1; SCDOS; Stearoyl CoA desaturase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6379