Learn More
Biomatik Corporation Mouse XOD (Xantine oxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF59195
Encompass_Preferred
Mouse XOD (Xantine oxidase) ELISA Kit for detecting XOD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q00519. Target Synonyms: XOD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6382