Learn More
Biomatik Corporation Rat OSM (Oncostatin-M) ELISA Kit
Supplier:  Biomatik Corporation EKF59201
Encompass_Preferred
Rat OSM (Oncostatin-M) ELISA Kit for detecting OSM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q65Z15. Target Synonyms: OSM; MGC20461. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6387