Learn More
Biomatik Corporation Mouse Gip (Gastric inhibitory polypeptide) ELISA Kit
Supplier:  Biomatik Corporation EKF59212
Encompass_Preferred
Mouse Gip (Gastric inhibitory polypeptide) ELISA Kit for detecting Gip in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48756. Target Synonyms: Gip; ; gastric inhibitory polypeptide; Glucose-dependent Insulinotropic Polypeptide; Incretin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6395