Learn More
Biomatik Corporation Human ENPP1 (Ectonucleotide pyrophosphatase/phosphodiesterase family member 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59219
Encompass_Preferred
Human ENPP1 (Ectonucleotide pyrophosphatase/phosphodiesterase family member 1) ELISA Kit for detecting ENPP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22413. Target Synonyms: ENPP1; Ly-41 Antigen; M6S1; NPP1; NPPS; PCA1; PDNP1; E-NPP 1; PC1; PC-1; ARHR2; Membrane component chromosome 6 surface marker 1; alkaline phosphodiesterase 1; plasma-cell membrane glycoprotein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6400