Learn More
Biomatik Corporation Human ZNRF2 (Zinc and ring finger 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59363
Encompass_Preferred
Human ZNRF2 (Zinc and ring finger 2) ELISA Kit for detecting ZNRF2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NHG8. Target Synonyms: Protein Ells2; RING finger protein 202; RNF202; zinc and ring finger 2; Zinc; RING finger protein 2; ZNRF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6408