Learn More
Biomatik Corporation Hamster Lep (Leptin) ELISA Kit
Supplier:  Biomatik Corporation EKF59548
Encompass_Preferred
Hamster Lep (Leptin) ELISA Kit for detecting Lep in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A1U7QZV4. Target Synonyms: Leptin; LEP; LEPD; OBS; Obesity Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6414