Learn More
Biomatik Corporation Rat IL-12/P70 (Interleukin 12 (p70)) ELISA Kit
Supplier:  Biomatik Corporation EKF59749
Encompass_Preferred
Rat IL-12/P70 (Interleukin 12 (p70)) ELISA Kit for detecting IL-12; P70 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R103. Target Synonyms: Interleukin 12; p70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6426