Learn More
Biomatik Corporation Human RIPK3 (Receptor-interacting serine/threonine-protein kinase 3) ELISA Kit
Supplier:  Biomatik Corporation EKF59931
Encompass_Preferred
Human RIPK3 (Receptor-interacting serine/threonine-protein kinase 3) ELISA Kit for detecting RIPK3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Y572. Target Synonyms: Ripk3; RIP3; Receptor-interacting protein 3; receptor-interacting serine-threonine kinase 3; RIP-3; RIP3receptor interacting protein 3; RIP-like protein kinase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6437