Learn More
Biomatik Corporation Human STK31 (Serine/threonine-protein kinase 31) ELISA Kit
Supplier:  Biomatik Corporation EKF59964
Encompass_Preferred
Human STK31 (Serine/threonine-protein kinase 31) ELISA Kit for detecting STK31 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9BXU1. Target Synonyms: serine; threonine kinase 31; SGK396; STK31; Sugen kinase 396; TDRD8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6440