Learn More
Biomatik Corporation Rat IL-12A (Interleukin 12 p35) ELISA Kit
Supplier:  Biomatik Corporation EKF60009
Encompass_Preferred
Rat IL-12A (Interleukin 12 p35) ELISA Kit for detecting IL-12A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R103. Target Synonyms: Interleukin 12 p35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6443