Learn More
Biomatik Corporation Human NEGR1 (Neuronal growth regulator 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60076
Encompass_Preferred
Human NEGR1 (Neuronal growth regulator 1) ELISA Kit for detecting NEGR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7Z3B1. Target Synonyms: NEGR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6445