Learn More
Biomatik Corporation Mouse MDA (Malonaldehyde) ELISA Kit
Supplier:  Biomatik Corporation EKF60157
Encompass_Preferred
Mouse MDA (Malonaldehyde) ELISA Kit for detecting MDA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: MDA; Malondialdehyde. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6452