Learn More
Biomatik Corporation Human IL-17 A/F (Interleukin 17A/F ) ELISA Kit
Supplier:  Biomatik Corporation EKF60392
Encompass_Preferred
Human IL-17 A/F (Interleukin 17A/F ) ELISA Kit for detecting IL-17 A; F in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q16552; Q96PD4. Target Synonyms: IL-17 A/F; Interleukin 17A/F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6462