Learn More
Biomatik Corporation Rat OCLN (Occludin) ELISA Kit
Supplier:  Biomatik Corporation EKF60414
Encompass_Preferred
Rat OCLN (Occludin) ELISA Kit for detecting OCLN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6P6T5. Target Synonyms: OCLN; BLCPMG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6468