Learn More
Biomatik Corporation Human CDK12 (Cyclin-dependent kinase 12) ELISA Kit
Supplier:  Biomatik Corporation EKF60420
Encompass_Preferred
Human CDK12 (Cyclin-dependent kinase 12) ELISA Kit for detecting CDK12 in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NYV4. Target Synonyms: CDK12; Cell division protein kinase 12; hCDK12; Cdc2-related kinase; arginine; serine-rich; CrkRS; Cell division cycle 2-related protein kinase 7; CDC2-related protein kinase 7;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6472