Learn More
Biomatik Corporation Human PGRP-S (Peptidoglycan Recognition Protein-S) ELISA Kit
Supplier:  Biomatik Corporation EKF60428
Encompass_Preferred
Human PGRP-S (Peptidoglycan Recognition Protein-S) ELISA Kit for detecting PGRP-S in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O75594. Target Synonyms: PGRP-S; TAG7; TNFSF3L; peptidoglycan recognition protein 1; Peptidoglycan recognition protein short; PGLYRP; PGRPpeptidoglycan recognition protein; PGRP-SMGC126894; PGRPSMGC126896; TAG7; TNF superfamily; member 3; LTB-like; peptidoglycan recognition protein; TNFSF3L; PGLYRP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6477