Learn More
Biomatik Corporation Rat Hepc 25 (Hepcidin 25) ELISA Kit
Supplier:  Biomatik Corporation EKF60439
Encompass_Preferred
Rat Hepc 25 (Hepcidin 25) ELISA Kit for detecting Hepc 25 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.5ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99MH3. Target Synonyms: Hepcidin 25. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6485