Learn More
Biomatik Corporation Mouse SYT1 (Synaptotagmin 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60441
Encompass_Preferred
Mouse SYT1 (Synaptotagmin 1) ELISA Kit for detecting SYT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P46096. Target Synonyms: SYT1; P65; SVP65; SYT1; DKFZp781D2042; p65; P65; SVP65; synaptotagmin Isynaptotagmin 1; synaptotagmin-1; SYT; SytI; sytI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6487