Learn More
Biomatik Corporation Human U-II (Urotensin 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60469
Encompass_Preferred
Human U-II (Urotensin 2) ELISA Kit for detecting U-II in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95399. Target Synonyms: U-II; Urotensin 2; UII; Urotensin II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6501