Learn More
Biomatik Corporation Human PTGS1 (Prostaglandin G/H synthase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60473
Encompass_Preferred
Human PTGS1 (Prostaglandin G/H synthase 1) ELISA Kit for detecting PTGS1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23219. Target Synonyms: PTGS1; COX-1; COX1; PGG; HS; COX3; PTGHS; PCOX1; PGH synthase 1; PGHS1; PGHS-1; PHS1; PHS 1; Cyclooxygenase-1; prostaglandin G; H synthase 1; prostaglandin G; H synthase and cyclooxygenase; Prostaglandin H2 synthase 1; Prostaglandin-endoperoxide synthase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6504