Learn More
Biomatik Corporation Mouse IL-23 (Interleukin 23) ELISA Kit
Supplier:  Biomatik Corporation EKF60476
Encompass_Preferred
Mouse IL-23 (Interleukin 23) ELISA Kit for detecting IL-23 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9EQ14. Target Synonyms: IL-23; IL-23A; IL23P19; P19; SGRF; IL23A; SGRF; IL-23; IL-23A; IL-23-A; IL-23p19; interleukin 23 p19 subunit; interleukin 23; alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; JKA3 induced upon T-cell activation; SGRFIL-23 subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6506