Learn More
Biomatik Corporation Mouse TIMP-4 (Tissue Inhibitors of Metalloproteinase 4) ELISA Kit
Supplier:  Biomatik Corporation EKF60484
Encompass_Preferred
Mouse TIMP-4 (Tissue Inhibitors of Metalloproteinase 4) ELISA Kit for detecting TIMP-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JHB3. Target Synonyms: TIMP-4; TIMP metallopeptidase inhibitor 4; tissue inhibitor of metalloproteinase 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6512