Learn More
Biomatik Corporation Mouse TGF-β2 (Transforming Growth Factor β2) ELISA Kit
Supplier:  Biomatik Corporation EKF60502
Encompass_Preferred
Mouse TGF-β2 (Transforming Growth Factor β2) ELISA Kit for detecting TGF-β2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27090. Target Synonyms: TGF-β2; Transforming Growth Factor β2; TGF-β2; TGFβ2; TGFB2; BSC-1 cell growth inhibitor; Cetermin; G-TSF; Polyergin; polyergin; TGF-beta2; TGF-beta-2; transforming growth factor beta-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6528