Learn More
Biomatik Corporation Mouse Ang2 (Angiopoietin-2) ELISA Kit
Supplier:  Biomatik Corporation EKF60512
Encompass_Preferred
Mouse Ang2 (Angiopoietin-2) ELISA Kit for detecting Ang2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.063-2500pg/ml. Sensitivity: 23.438pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q64438. Target Synonyms: ANG2; Angiopoietin 2; ANG-2; ANGPT2; AGPT2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6538