Learn More
Biomatik Corporation Mouse BMP-7 (Bone morphogenetic protein 7) ELISA Kit
Supplier:  Biomatik Corporation EKF60519
Encompass_Preferred
Mouse BMP-7 (Bone morphogenetic protein 7) ELISA Kit for detecting BMP-7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23359. Target Synonyms: BMP-7; Bone Morphogenetic Protein 7; ; BMP7; OP1; Osteogenic Protein-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6545