Learn More
Biomatik Corporation Mouse GROβ (Growth Regulated Oncogene Beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60536
Encompass_Preferred
Mouse GROβ (Growth Regulated Oncogene Beta) ELISA Kit for detecting GROβ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10889. Target Synonyms: GROβ; CXCL2; MIP-2a; CINC-3; GRO betaGRO2; GRO2 oncogene; GROb; GROB; CINC-2a; MGSA beta; MGSA-b; MIP2A; MIP2-alpha; SCYB2; MGSA-beta; chemokine; C-X-C motif ligand 2; C-X-C Motif Chemokine 2; Gro-beta; gro-beta; Growth-regulated protein beta; Macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6559