Learn More
Biomatik Corporation Human Mast Cell Chymase ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34542
Encompass_Preferred
The Human Mast Cell Chymase ELISA Kit allows for quantitative determination of chymase 1, mast cell in serum, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P23946. Restriction: For Research Use Only.
Catalog No. 50-149-8059