Learn More
Biomatik Corporation Rat TPH1 (Tryptophan Hydroxylase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60563
Encompass_Preferred
Rat TPH1 (Tryptophan Hydroxylase 1) ELISA Kit for detecting TPH1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09810. Target Synonyms: TPH1; TPRH; indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase; tryptophan 5-monooxygenase; tryptophan hydroxylase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6586