Learn More
Biomatik Corporation Human FASL (Factor Related Apoptosis Ligand) ELISA Kit
Supplier:  Biomatik Corporation EKF60575
Encompass_Preferred
Human FASL (Factor Related Apoptosis Ligand) ELISA Kit for detecting FASL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48023. Target Synonyms: FASL; TNFSF6; FASLG; ALPS1B; APT1LG1; APTL; CD178; CD95L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6598