Learn More
Biomatik Corporation Rat MDC/CCL22 (Macrophage-Derived Chemokine) ELISA Kit
Supplier:  Biomatik Corporation EKF60580
Encompass_Preferred
Rat MDC/CCL22 (Macrophage-Derived Chemokine) ELISA Kit for detecting MDC; CCL22 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QZU1. Target Synonyms: MDC; CCL22; ABCD-1; MDC; STCP-1; A-152E5.1; ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; chemokine; C-C motif ligand 22; DC; B-CK; Macrophage-derived chemokine; MDCStimulated T-cell chemotactic protein 1; MGC34554; SCYA22MDC; 1-69; small inducible cytokine A22; small inducible cytokine subfamily A; Cys-Cys; member 22; Small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6603