Learn More
Biomatik Corporation Rat Bax (Apoptosis regulator BAX) ELISA Kit
Supplier:  Biomatik Corporation EKF60582
Encompass_Preferred
Rat Bax (Apoptosis regulator BAX) ELISA Kit for detecting Bax in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63690. Target Synonyms: Bax; apoptosis regulator BAX; BCL2-associated X protein; Bcl2-L-4; BCL2L4bcl2-L-4; Bcl-2-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6605