Learn More
Biomatik Corporation Human OGN (Mimecan) ELISA Kit
Supplier:  Biomatik Corporation EKF60588
Encompass_Preferred
Human OGN (Mimecan) ELISA Kit for detecting OGN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20774. Target Synonyms: OGN; Osteoglycin; Osteoinductive factor; KSPG25; OIF; SLRR3A; DKFZp586P2421; mimecan; mimecan proteoglycan; OIFOG; Osteoglycin; osteoglycin; osteoglycin; osteoinductive factor; Osteoinductive factor; SLRR3Acorneal keRatan sulfate proteoglycan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6611