Learn More
Biomatik Corporation Human NT-ProANP (N-Terminal Pro Atrial Natriuretic Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF60594
Encompass_Preferred
Human NT-ProANP (N-Terminal Pro Atrial Natriuretic Peptide) ELISA Kit for detecting NT-ProANP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01160. Target Synonyms: NT-ProANP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6617