Learn More
Biomatik Corporation Mouse S100B (S100 Calcium Binding Protein B) ELISA Kit
Supplier:  Biomatik Corporation EKF60597
Encompass_Preferred
Mouse S100B (S100 Calcium Binding Protein B) ELISA Kit for detecting S100B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50114. Target Synonyms: S100B; Protein S100-B; S100; S100 beta; S100 calcium binding protein B; S100 calcium-binding protein B; S100 calcium-binding protein; beta; neural; S-100 calcium-binding protein; beta chain; 10; S-100 protein beta chain; S-100 protein subunit beta; S100beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6620