Learn More
Biomatik Corporation Human LPO (Lactoperoxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF60601
Encompass_Preferred
Human LPO (Lactoperoxidase) ELISA Kit for detecting LPO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22079. Target Synonyms: LPO; Lactoperoxidase; Salivary peroxidase; SPO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6624