Learn More
Biomatik Corporation Rat St2a1 (Bile salt sulfotransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF60655
Encompass_Preferred
Rat St2a1 (Bile salt sulfotransferase) ELISA Kit for detecting St2a1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15709. Target Synonyms: St2a1; DHEAS; DHEA-ST; hSTa; ST2A3; SULT2A1; alcohol; hydroxysteroid sulfotransferase; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-ST; DHEA-STbile salt sulfotransferase; EC 2.8.2.14; HST; hSTa; Hydroxysteroid Sulfotransferase; ST2; ST2A1; ST2A3; STDbile-salt sulfotranasferase 2A1; Sulfotransferase 2A1; sulfotransferase family; cytosolic; 2A; dehydroepiandrosterone; DHEA-preferring; member 1; SULT2A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6672