Learn More
Biomatik Corporation Rat GSK3β (Glycogen synthase kinase-3 beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60666
Encompass_Preferred
Rat GSK3β (Glycogen synthase kinase-3 beta) ELISA Kit for detecting GSK3β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18266. Target Synonyms: GSK3B; GSK-3 beta; Serine; threonine-protein kinase GSK3B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6681