Learn More
Biomatik Corporation Human SERPINB4 (Serpin B4) ELISA Kit
Supplier:  Biomatik Corporation EKF60669
Encompass_Preferred
Human SERPINB4 (Serpin B4) ELISA Kit for detecting SERPINB4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48594. Target Synonyms: SERPINB4; PI11; SCCA2; Serpin B4; Leupin; Peptidase inhibitor 11; PI-11; Squamous cell carcinoma antigen 2; SCCA-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6684