Learn More
Biomatik Corporation Rat CCK-8 (Cholecystokinin 8) ELISA Kit
Supplier:  Biomatik Corporation EKF60675
Encompass_Preferred
Rat CCK-8 (Cholecystokinin 8) ELISA Kit for detecting CCK-8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01355. Target Synonyms: CCK-8; Cholecystokinin 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6689