Learn More
Biomatik Corporation Human TIE1 (Tyrosine-protein kinase receptor Tie-1) ELISA Kit
Supplier:  Biomatik Corporation EKF60680
Encompass_Preferred
Human TIE1 (Tyrosine-protein kinase receptor Tie-1) ELISA Kit for detecting TIE1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35590. Target Synonyms: TIE1; Tyrosine-protein kinase receptor Tie-1; receptor tyrosine kinase; soluble TIE1 variant 1; soluble TIE1 variant 2; soluble TIE1 variant 5; TIEsoluble TIE1 variant 4; tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6692