Learn More
Biomatik Corporation Human GSK3β (Glycogen synthase kinase-3 beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60692
Encompass_Preferred
Human GSK3β (Glycogen synthase kinase-3 beta) ELISA Kit for detecting GSK3β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49841. Target Synonyms: GSK3β; Glycogen Synthase Kinase 3 Beta; GSK3B; glycogen synthase kinase-3 beta; GSK-3 beta; GSK3beta isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6704