Learn More
Biomatik Corporation Porcine cTn-I/TNNI3 (Cardiac Troponin I) ELISA Kit
Supplier:  Biomatik Corporation EKF60715
Encompass_Preferred
Porcine cTn-I/TNNI3 (Cardiac Troponin I) ELISA Kit for detecting cTn-I; TNNI3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A5X5T5. Target Synonyms: cTnI; TNNC1; CMD1FF; CMD2A; CMH7; RCM1; cTn-I; TNNI3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6711