Learn More
Biomatik Corporation Porcine VEGFR1/Flt1 (Vascuoar Endothelial Growth Factor Receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60724
Encompass_Preferred
Porcine VEGFR1/Flt1 (Vascuoar Endothelial Growth Factor Receptor 1) ELISA Kit for detecting VEGFR1; Flt1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: F1RSU5. Target Synonyms: VEGFR1; FLT1; Vascular Endothelial Growth Factor Receptor 1; FLT1; Flt-1; FLT-1; Fms-like tyrosine kinase 1; fms-related tyrosine kinase 1; vascular endothelial growth factor; vascularpermeability factor receptor; FRT; Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; vascular endothelial growth factor receptor 1; Vascular permeability factor receptor; VEGFR1; VEGFR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6714