Learn More
Biomatik Corporation Human ICAM-3 (Intercellular adhesion molecule 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60734
Encompass_Preferred
Human ICAM-3 (Intercellular adhesion molecule 3) ELISA Kit for detecting ICAM-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P32942. Target Synonyms: ICAM-3; CD50. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6716