Learn More
Biomatik Corporation Mouse GAD-Ab-IgM (Glutamic Acid Decarboxylase Autoantibody IgM) ELISA Kit
Supplier:  Biomatik Corporation EKF60776
Encompass_Preferred
Mouse GAD-Ab-IgM (Glutamic Acid Decarboxylase Autoantibody IgM) ELISA Kit for detecting GAD-Ab-IgM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Indirect ELISA. Assay Time: 3h, 30 min. Accession Number: P48320. Target Synonyms: GAD-Ab-IgM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6731